Learn More
Medchemexpress LLC Albendazole sulfoxide | 54029-12-8 | 99.2% | 1 ML
SDP
Catalog No. 5000339754
Encompass_Preferred
Albendazole sulfoxide, also known as Ricobendazole, is the primary active metabolite of Albendazole. It exhibits anti-parasitic effects against *Echinococcus multilocularis* Metacestodes, demonstrating parasitocidal activity in vitro and reducing cyst size in vivo.
  • Main active metabolite of Albendazole
  • Exhibits anti-parasitic effects
  • Active against *Echinococcus multilocularis* Metacestodes
  • Demonstrates parasitocidal effects in vitro
  • Reduces cyst size and weight in vivo
Catalog No. 50-003-39754 Supplier Medchemexpress LLC Supplier No. HY1278510MM/1ML
Albendazole sulfoxide, also known as Ricobendazole, is the primary active metabolite of Albendazole. It exhibits anti-parasitic effects against *Echinococcus multilocularis* Metacestodes, demonstrating parasitocidal activity in vitro and reducing cyst size in vivo.
  • Main active metabolite of Albendazole
  • Exhibits anti-parasitic effects
  • Active against *Echinococcus multilocularis* Metacestodes
  • Demonstrates parasitocidal effects in vitro
  • Reduces cyst size and weight in vivo