Learn More
Medchemexpress LLC Albendazole sulfoxide | 54029-12-8 | 99.2% | 100 MG
SDP
Catalog No. 5000341674
Encompass_Preferred
Albendazole sulfoxide (Ricobendazole) is the main active metabolite of Albendazole, demonstrating an anti-parasite effect against *Echinococcus multilocularis* Metacestodes. It is intended for research use only and not for sale to patients. This compound is a solid, white to off-white in appearance, with a molecular weight of 281.33.
  • Exhibits a parasitocidal effect against *E. multilocularis* metacestodes in vitro.
  • Reduces cyst size and weight in Balb/c mice infected with protoscoleces in vivo.
  • Available in various solid package sizes and as a solution in DMSO.
  • Stable under recommended storage conditions, with specific storage instructions for powder and in solvent.
  • For research use only by suitably qualified scientists.
Catalog No. 50-003-41674 Supplier Medchemexpress LLC Supplier No. HY12785100MG
Albendazole sulfoxide (Ricobendazole) is the main active metabolite of Albendazole, demonstrating an anti-parasite effect against *Echinococcus multilocularis* Metacestodes. It is intended for research use only and not for sale to patients. This compound is a solid, white to off-white in appearance, with a molecular weight of 281.33.
  • Exhibits a parasitocidal effect against *E. multilocularis* metacestodes in vitro.
  • Reduces cyst size and weight in Balb/c mice infected with protoscoleces in vivo.
  • Available in various solid package sizes and as a solution in DMSO.
  • Stable under recommended storage conditions, with specific storage instructions for powder and in solvent.
  • For research use only by suitably qualified scientists.