Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Albendazole sulfoxide-d3 | 1448346-38-0 | MFCD26142908 | 99.8% | 284.35 | C12H12D3N3O3S | 5 MG
SDP

Catalog No. 5000264470
Encompass_Preferred

Albendazole sulfoxide-d3 is a deuterium-labeled analogue of albendazole sulfoxide supplied for research use. It is intended as an isotope-labeled tracer or internal standard for quantitative analyses by NMR, GC-MS, and LC-MS.

  • Deuterium-labeled internal standard for quantitative analysis.
  • Suitable for NMR, GC-MS, and LC-MS methods.
  • High purity (99.79%).
  • Molecular weight 284.35 and formula C12H12D3N3O3S.
  • Available in small mg-scale packages, including 5 mg.
  • Storage conditions for powder and solution provided for long-term stability.

Catalog No. 50-002-64470 Supplier Medchemexpress LLC Supplier No. HY12785S5MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Albendazole sulfoxide-d3 is a deuterium-labeled analogue of albendazole sulfoxide supplied for research use. It is intended as an isotope-labeled tracer or internal standard for quantitative analyses by NMR, GC-MS, and LC-MS.

  • Deuterium-labeled internal standard for quantitative analysis.
  • Suitable for NMR, GC-MS, and LC-MS methods.
  • High purity (99.79%).
  • Molecular weight 284.35 and formula C12H12D3N3O3S.
  • Available in small mg-scale packages, including 5 mg.
  • Storage conditions for powder and solution provided for long-term stability.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.