Learn More
MSC Albion Caster, 4 inch, Soft Rubber, 115 Lb, Friction Stem
Catalog No. 5000529905
Encompass
This caster is a stem mount swivel type, featuring a 4 inch diameter soft rubber wheel. It is designed for various applications including appliances, equipment, and furniture, offering a load capacity of 115 Lb. The caster includes a self-lubricating bearing for smooth operation and a friction stem for secure mounting.
  • 4 inch wheel diameter
  • Soft rubber wheel material
  • 115 Lb load capacity
  • Self-lubricating bearing for smooth operation
  • Friction stem swivel type for secure mounting
  • Suitable for appliances, equipment, and furniture
  • Black wheel color
  • Zinc finish
Catalog No. 50-005-29905 Supplier MSC Supplier No. 00042465
This caster is a stem mount swivel type, featuring a 4 inch diameter soft rubber wheel. It is designed for various applications including appliances, equipment, and furniture, offering a load capacity of 115 Lb. The caster includes a self-lubricating bearing for smooth operation and a friction stem for secure mounting.
  • 4 inch wheel diameter
  • Soft rubber wheel material
  • 115 Lb load capacity
  • Self-lubricating bearing for smooth operation
  • Friction stem swivel type for secure mounting
  • Suitable for appliances, equipment, and furniture
  • Black wheel color
  • Zinc finish