Learn More
MSC Albion Caster, 72 Series, Swivel, Phenolic, 6 in x 2 in
Catalog No. 5000527531
Encompass
These 72 Series casters are constructed from hot-forged steel, ensuring extended life in rugged applications. Designed to outperform cold-forged or stamped steel casters in swivel applications, they are ideal for production racks, fixtures, and box trucks. They feature larger top plates and higher load capacities compared to the 71 Series, with a Zerk fitting in the swivel raceway for easy maintenance.
  • Swivel caster type
  • Phenolic wheel material
  • 6 in wheel diameter
  • 2 in tread width
  • Roller wheel bearing
  • Top plate mounting type
  • 1,200 lbs load capacity
  • 7-1/2 in mounting height
  • 4-1/2 in x 6-1/4 in plate dimensions
  • Zinc finish
  • Hot forged construction
  • Zerk fitting for easy maintenance
Catalog No. 50-005-27531 Supplier MSC Supplier No. 75948745
These 72 Series casters are constructed from hot-forged steel, ensuring extended life in rugged applications. Designed to outperform cold-forged or stamped steel casters in swivel applications, they are ideal for production racks, fixtures, and box trucks. They feature larger top plates and higher load capacities compared to the 71 Series, with a Zerk fitting in the swivel raceway for easy maintenance.
  • Swivel caster type
  • Phenolic wheel material
  • 6 in wheel diameter
  • 2 in tread width
  • Roller wheel bearing
  • Top plate mounting type
  • 1,200 lbs load capacity
  • 7-1/2 in mounting height
  • 4-1/2 in x 6-1/4 in plate dimensions
  • Zinc finish
  • Hot forged construction
  • Zerk fitting for easy maintenance