Learn More
MSC Albion Caster Grip Lock Brake Kit, 4 inch diameter, Face Contact, Pedal Trailing
Catalog No. 5000533383
Encompass
This Albion Caster Grip Lock Brake Kit is designed for use with 4 inch casters. It is a face contact brake with a trailing pedal, compatible with the 16 series casters. This brake option does not work with swivel locks.
  • Designed for 4 inch casters
  • Grip lock brake mechanism
  • Features a face contact design
  • Equipped with a trailing pedal
  • Compatible with 16 series casters
  • Does not integrate with swivel locks
  • Supplied as a four piece kit
Catalog No. 50-005-33383 Supplier MSC Supplier No. 01859057
This Albion Caster Grip Lock Brake Kit is designed for use with 4 inch casters. It is a face contact brake with a trailing pedal, compatible with the 16 series casters. This brake option does not work with swivel locks.
  • Designed for 4 inch casters
  • Grip lock brake mechanism
  • Features a face contact design
  • Equipped with a trailing pedal
  • Compatible with 16 series casters
  • Does not integrate with swivel locks
  • Supplied as a four piece kit