Learn More
MSC Albion Caster, Rigid, 8 inch Wheel Diameter, 2 inch Tread Width, Moldon Rubber, 600 lb Capacity
Catalog No. 5000537554
Encompass
The Albion 72 Series Rigid Caster is engineered for demanding applications, featuring hot-forged steel construction for extended durability. This caster provides reliable performance and easy maintenance, making it suitable for various industrial material handling environments.
  • Constructed from hot-forged steel for extended durability
  • Features a Zerk fitting in the swivel raceway for easy lubrication
  • Designed with larger top plates and higher load capacities for heavy-duty use
  • Equipped with a moldon rubber wheel on cast iron for smooth operation
  • Offers a load capacity of 600 lbs
  • Includes 1/4 inch steel legs and a hollow axle
  • Has an overall height of 10-1/8 inch
  • Finished with a protective zinc coating
Catalog No. 50-005-37554 Supplier MSC Supplier No. 75948711
The Albion 72 Series Rigid Caster is engineered for demanding applications, featuring hot-forged steel construction for extended durability. This caster provides reliable performance and easy maintenance, making it suitable for various industrial material handling environments.
  • Constructed from hot-forged steel for extended durability
  • Features a Zerk fitting in the swivel raceway for easy lubrication
  • Designed with larger top plates and higher load capacities for heavy-duty use
  • Equipped with a moldon rubber wheel on cast iron for smooth operation
  • Offers a load capacity of 600 lbs
  • Includes 1/4 inch steel legs and a hollow axle
  • Has an overall height of 10-1/8 inch
  • Finished with a protective zinc coating