Learn More
MSC Albion Caster, Rigid, Glass-Filled Nylon, 4 in Wheel Diameter, 800 lb Capacity
Catalog No. 5000534601
Encompass
This rigid top plate caster features a glass-filled nylon wheel with a 4 inch diameter and 2 inch width, providing an 800 lb capacity and an overall height of 5-5/8 inches. It is designed for ergonomic movement and quiet operation, featuring a precision sealed bearing for maintenance-free use and reduced push/pull force. The rig is zinc-plated for corrosion resistance.
  • Maintenance-free operation
  • Zinc-plated finish for corrosion resistance
  • Precision sealed ball bearing
  • Equipped with a 1/2 inch diameter solid axle
  • Features 1/4 inch steel legs
  • Designed for ergonomic movement and quiet mobility
  • Reduces push/pull force
Catalog No. 50-005-34601 Supplier MSC Supplier No. 74583022
This rigid top plate caster features a glass-filled nylon wheel with a 4 inch diameter and 2 inch width, providing an 800 lb capacity and an overall height of 5-5/8 inches. It is designed for ergonomic movement and quiet operation, featuring a precision sealed bearing for maintenance-free use and reduced push/pull force. The rig is zinc-plated for corrosion resistance.
  • Maintenance-free operation
  • Zinc-plated finish for corrosion resistance
  • Precision sealed ball bearing
  • Equipped with a 1/2 inch diameter solid axle
  • Features 1/4 inch steel legs
  • Designed for ergonomic movement and quiet mobility
  • Reduces push/pull force