Learn More
MSC Albion Caster, Rigid, Polyurethane, 5 in Wheel Diameter, 350 lb Capacity
Catalog No. 5000512979
Encompass
This caster is a rigid top plate model designed for reliable performance in material handling applications. It features a durable polyurethane wheel with a 5-inch diameter, capable of supporting a load of up to 350 pounds, making it suitable for various industrial and commercial settings.
  • Polyurethane wheel material for durability
  • 5 inch wheel diameter
  • 1-1/4 inch wheel width
  • 350 lb load capacity
  • Plate mounting for secure attachment
  • Overall height of 6-3/16 inches
  • Zinc finish to resist rust and corrosion
  • Equipped with a 3/8 in diameter hollow axle with lock nut
  • Suitable for material handling applications
Catalog No. 50-005-12979 Supplier MSC Supplier No. 75947135
This caster is a rigid top plate model designed for reliable performance in material handling applications. It features a durable polyurethane wheel with a 5-inch diameter, capable of supporting a load of up to 350 pounds, making it suitable for various industrial and commercial settings.
  • Polyurethane wheel material for durability
  • 5 inch wheel diameter
  • 1-1/4 inch wheel width
  • 350 lb load capacity
  • Plate mounting for secure attachment
  • Overall height of 6-3/16 inches
  • Zinc finish to resist rust and corrosion
  • Equipped with a 3/8 in diameter hollow axle with lock nut
  • Suitable for material handling applications