Learn More
MSC Albion Caster, Rigid Top Plate, Polyurethane, 4 in Wheel, 350 lb Capacity
Catalog No. 5000531631
Encompass
This rigid top plate caster features a durable polyurethane wheel with a 4-inch diameter and a 350 lb load capacity. It is designed for applications requiring fixed direction movement and reliable performance.
  • Rigid top plate design
  • Polyurethane wheel material
  • 4 in wheel diameter
  • 1-1/4 in wheel width
  • 350 lb load capacity
  • 5-1/8 in overall height
  • 3.625 in plate length
  • 2.5 in plate width
Catalog No. 50-005-31631 Supplier MSC Supplier No. 75947101
This rigid top plate caster features a durable polyurethane wheel with a 4-inch diameter and a 350 lb load capacity. It is designed for applications requiring fixed direction movement and reliable performance.
  • Rigid top plate design
  • Polyurethane wheel material
  • 4 in wheel diameter
  • 1-1/4 in wheel width
  • 350 lb load capacity
  • 5-1/8 in overall height
  • 3.625 in plate length
  • 2.5 in plate width