Learn More
MSC Albion Caster, Rigid Top Plate, Soft Rubber, 6 in Wheel, 600 lb
Catalog No. 5000534754
Encompass
This rigid top plate caster is designed for material handling applications, featuring a soft rubber wheel with a 6 in diameter and 2 in width. It has a load capacity of 600 lb, a zinc finish, and a cold forged construction for durability.
  • Features a 1/2 in diameter solid axle and 1/4 in steel legs.
  • Designed to be maintenance free.
  • Includes a top plate and yoke base.
  • Equipped with a precision sealed ball bearing.
  • Comes with a sealed wheel bearing type.
Catalog No. 50-005-34754 Supplier MSC Supplier No. 74877648
This rigid top plate caster is designed for material handling applications, featuring a soft rubber wheel with a 6 in diameter and 2 in width. It has a load capacity of 600 lb, a zinc finish, and a cold forged construction for durability.
  • Features a 1/2 in diameter solid axle and 1/4 in steel legs.
  • Designed to be maintenance free.
  • Includes a top plate and yoke base.
  • Equipped with a precision sealed ball bearing.
  • Comes with a sealed wheel bearing type.