Learn More
MSC Albion Caster, Swivel, Glass-Filled Nylon, 6 in, 1200 lb Capacity
Catalog No. 5000527827
Encompass
This industrial caster features a swivel top plate and a durable glass-filled nylon wheel. Designed for material handling applications, it offers a 1200 lb capacity and a 6 inch wheel diameter, ensuring reliable performance.
  • Swivel section
  • 1/2 in diameter solid axle
  • 1/4 in steel legs
  • Maintenance free
  • Top plate and yoke base
Catalog No. 50-005-27827 Supplier MSC Supplier No. 74583113
This industrial caster features a swivel top plate and a durable glass-filled nylon wheel. Designed for material handling applications, it offers a 1200 lb capacity and a 6 inch wheel diameter, ensuring reliable performance.
  • Swivel section
  • 1/2 in diameter solid axle
  • 1/4 in steel legs
  • Maintenance free
  • Top plate and yoke base