Learn More
MSC Albion Caster Wheel, Cast Iron, 3-1/4 in Diameter, Roller Bearing
Catalog No. 5000533517
Encompass
This caster wheel is a durable cast iron wheel designed for high-speed and high-capacity applications. Featuring a 3-1/4 in diameter and a straight roller bearing, it offers exceptional performance and can withstand temperatures up to +800°F. This robust wheel provides a cost-effective solution for demanding environments.
  • Heat resistant
  • Impact resistant
  • Premium quality
  • Cast iron core
  • High capacity
  • Steel replacement
  • Flat tread
Catalog No. 50-005-33517 Supplier MSC Supplier No. 00996538
This caster wheel is a durable cast iron wheel designed for high-speed and high-capacity applications. Featuring a 3-1/4 in diameter and a straight roller bearing, it offers exceptional performance and can withstand temperatures up to +800°F. This robust wheel provides a cost-effective solution for demanding environments.
  • Heat resistant
  • Impact resistant
  • Premium quality
  • Cast iron core
  • High capacity
  • Steel replacement
  • Flat tread