Learn More
MSC Albion Caster Wheel, Phenolic, 3-1/2 in, 1-1/4 in, Plain Bearing
Catalog No. 5000529670
Encompass
This caster wheel features a phenolic wheel material with a 3-1/2 inch diameter and 1-1/4 inch width. It incorporates a plain bearing for efficient movement and is built to withstand temperatures from -65°F to 250°F. The wheel is non-marking, has a plastic core, and comes in black.
  • Phenolic wheel material for durability.
  • Non-marking design protects floors.
  • Wide operating temperature range from -65°F to 250°F.
  • Plastic wheel core.
  • Black wheel color.
  • Plain bearing for smooth and reliable operation.
Catalog No. 50-005-29670 Supplier MSC Supplier No. 00995696
This caster wheel features a phenolic wheel material with a 3-1/2 inch diameter and 1-1/4 inch width. It incorporates a plain bearing for efficient movement and is built to withstand temperatures from -65°F to 250°F. The wheel is non-marking, has a plastic core, and comes in black.
  • Phenolic wheel material for durability.
  • Non-marking design protects floors.
  • Wide operating temperature range from -65°F to 250°F.
  • Plastic wheel core.
  • Black wheel color.
  • Plain bearing for smooth and reliable operation.