Learn More
MSC Albion Caster Wheel, Phenolic, 3 in Dia, 1-13/16 in Wide, Roller Bearing
Catalog No. 5000530811
Encompass
This caster wheel features a phenolic wheel with a 3 inch diameter and 1-13/16 inch width. It is designed with a 3/8 inch axle and a roller bearing, suitable for various applications. The wheel core is made of plastic, and it operates efficiently in temperatures from -65 °F to 250 °F.
  • Phenolic wheel material
  • 3 in diameter
  • 1-13/16 in width
  • 3/8 in axle size
  • Roller bearing type
  • Plastic wheel core material
  • Black wheel color
  • Operates in temperatures from -65 °F to 250 °F
  • Non-marking design
  • Manufactured in the United States
Catalog No. 50-005-30811 Supplier MSC Supplier No. 66043811
This caster wheel features a phenolic wheel with a 3 inch diameter and 1-13/16 inch width. It is designed with a 3/8 inch axle and a roller bearing, suitable for various applications. The wheel core is made of plastic, and it operates efficiently in temperatures from -65 °F to 250 °F.
  • Phenolic wheel material
  • 3 in diameter
  • 1-13/16 in width
  • 3/8 in axle size
  • Roller bearing type
  • Plastic wheel core material
  • Black wheel color
  • Operates in temperatures from -65 °F to 250 °F
  • Non-marking design
  • Manufactured in the United States