Learn More
MSC Albion Caster Wheel, Polyurethane, 4 in Dia, 1-1/2 in Wide, 1/2 in Axle
Catalog No. 5000513427
Encompass
This caster wheel is constructed from polyurethane, offering durability and smooth operation. It features a 4 inch diameter and 1-1/2 inch width, designed for various applications. The wheel includes a 1/2 inch axle and a roller bearing for efficient movement. It is non-marking and suitable for use in temperatures ranging from -40 F to 180 F.
  • Polyurethane wheel material
  • 4 inch diameter
  • 1-1/2 inch width
  • 1/2 inch axle size
  • Roller bearing type
  • Plastic wheel core material
  • Non-marking design
  • Wide operating temperature range from -40 F to 180 F
Catalog No. 50-005-13427 Supplier MSC Supplier No. 00995977
This caster wheel is constructed from polyurethane, offering durability and smooth operation. It features a 4 inch diameter and 1-1/2 inch width, designed for various applications. The wheel includes a 1/2 inch axle and a roller bearing for efficient movement. It is non-marking and suitable for use in temperatures ranging from -40 F to 180 F.
  • Polyurethane wheel material
  • 4 inch diameter
  • 1-1/2 inch width
  • 1/2 inch axle size
  • Roller bearing type
  • Plastic wheel core material
  • Non-marking design
  • Wide operating temperature range from -40 F to 180 F