Learn More
MSC Albion Casters Swivel Caster, 8 in Phenolic, 2500 Lb, Plate Mount
Catalog No. 5000513010
Encompass
This swivel caster features an 8 inch diameter phenolic wheel and a robust 2500 pound load capacity, making it suitable for heavy-duty material handling applications. Designed with a top plate mount, it provides reliable mobility and support.
  • Hollow axle
  • Steel plate legs
  • Swivel load bearing
  • Swivel top plate
  • Yoke base
  • Zerk fitting lubrication
Catalog No. 50-005-13010 Supplier MSC Supplier No. 75949172
This swivel caster features an 8 inch diameter phenolic wheel and a robust 2500 pound load capacity, making it suitable for heavy-duty material handling applications. Designed with a top plate mount, it provides reliable mobility and support.
  • Hollow axle
  • Steel plate legs
  • Swivel load bearing
  • Swivel top plate
  • Yoke base
  • Zerk fitting lubrication