Learn More
MSC Albion Floor Lock, 7-1/2 in Mounting Height, 6 in Caster Wheel Diameter
Catalog No. 5000512545
Encompass
This floor lock is designed to provide secure positioning for equipment featuring caster wheels, ensuring stability in various operational environments.
  • Features a 7-1/2 in mounting height
  • Compatible with 6 in diameter caster wheels
  • Provides 1-1/2 in retracted clearance
  • Extends to a height of 7.5 in
  • Finished in a durable powder coated yellow color
Catalog No. 50-005-12545 Supplier MSC Supplier No. 62632211
This floor lock is designed to provide secure positioning for equipment featuring caster wheels, ensuring stability in various operational environments.
  • Features a 7-1/2 in mounting height
  • Compatible with 6 in diameter caster wheels
  • Provides 1-1/2 in retracted clearance
  • Extends to a height of 7.5 in
  • Finished in a durable powder coated yellow color