Learn More
MSC Albion Rigid Top Plate Caster, Soft Rubber, 5 in Wheel Dia, 1-1/4 in Wheel Width, 175 lb Capacity
Catalog No. 5000531427
Encompass
The Albion 02RR05041R Rigid Top Plate Caster is made of soft rubber with a 5 in wheel diameter and 1-1/4 in wheel width. It has a 175 lb capacity and an overall height of 6-3/16 in.
  • Stamp formed construction.
  • Rigid style for stability.
  • Suitable for material handling applications.
  • Top plate mount for secure attachment.
  • Soft rubber wheel material for smooth operation.
  • Plain bearing type.
  • Black wheel color.
  • Features a 3/8 in diameter hollow axle with lock nut.
  • Single wheel design.
  • Zinc finish/coating for durability.
  • Overall height of 6-3/16 in.
  • Without wheel bearing.
  • Plate size of 2-1/2 x 3-5/8 in.
  • Soft wheel hardness.
Catalog No. 50-005-31427 Supplier MSC Supplier No. 75946798
The Albion 02RR05041R Rigid Top Plate Caster is made of soft rubber with a 5 in wheel diameter and 1-1/4 in wheel width. It has a 175 lb capacity and an overall height of 6-3/16 in.
  • Stamp formed construction.
  • Rigid style for stability.
  • Suitable for material handling applications.
  • Top plate mount for secure attachment.
  • Soft rubber wheel material for smooth operation.
  • Plain bearing type.
  • Black wheel color.
  • Features a 3/8 in diameter hollow axle with lock nut.
  • Single wheel design.
  • Zinc finish/coating for durability.
  • Overall height of 6-3/16 in.
  • Without wheel bearing.
  • Plate size of 2-1/2 x 3-5/8 in.
  • Soft wheel hardness.