Learn More
MSC Albion Swivel Caster, Cast Iron, 4 in Wheel, 700 lb Capacity
Catalog No. 5000527530
Encompass
This swivel top plate caster features a cast iron wheel with a 4-inch diameter and 2-inch width, offering a 700 lb load capacity and an overall height of 5-5/8 inches. It is designed for various industrial applications, providing reliable mobility and durability.
  • Cast iron wheel material for durability.
  • 4-inch wheel diameter.
  • 2-inch wheel width.
  • 700 lb load capacity.
  • Roller bearing for smooth operation.
  • Cold forged construction for strength.
  • Zinc finish for corrosion resistance.
  • Top plate mount for secure attachment.
  • Ideal for conveyor, dragline operations, mobile racks, and utility trucks.
Catalog No. 50-005-27530 Supplier MSC Supplier No. 75948646
This swivel top plate caster features a cast iron wheel with a 4-inch diameter and 2-inch width, offering a 700 lb load capacity and an overall height of 5-5/8 inches. It is designed for various industrial applications, providing reliable mobility and durability.
  • Cast iron wheel material for durability.
  • 4-inch wheel diameter.
  • 2-inch wheel width.
  • 700 lb load capacity.
  • Roller bearing for smooth operation.
  • Cold forged construction for strength.
  • Zinc finish for corrosion resistance.
  • Top plate mount for secure attachment.
  • Ideal for conveyor, dragline operations, mobile racks, and utility trucks.