The Albion 72 Series swivel casters are constructed from hot-forged steel, engineered for durability and extended use in demanding industrial environments. Designed to outperform cold-forged or stamped steel casters, they feature a zerk fitting in the swivel raceway for simplified maintenance. These casters are suitable for a wide range of heavy-duty applications, including production racks, box trucks, and various warehousing needs.
- Hot-forged steel construction for rugged environments.
- Zerk fitting in swivel raceway for easy maintenance.
- Suitable for production racks, fixtures, and box trucks.
- Larger top plates for enhanced stability.
- High load capacities for demanding applications.
- Multiple wheel diameter options: 4, 5, 6, 8, 10 inches.
- Crowned tread wheel face.
- Operating temperature range: -40°F to +180°F.
- Equipped with roller bearings.