Learn More
MSC Albion Swivel Caster, Phenolic, 6 inch Wheel, 1,200 lb Capacity
Catalog No. 5000527880
Encompass
This swivel top plate caster features a phenolic wheel with a 6 inch diameter, a 2 inch width, and a 1,200 lb load capacity. Manufactured from cold-formed steel, it includes a heat-treated king bolt and nut for durability. The rig is zinc-plated for corrosion resistance, and zerk fittings in the heat-treated swivel section allow for easy maintenance. It is suitable for material handling and impact-resistant applications.
  • Rolling ease
  • Floor protection
  • Quiet operation
  • Resistance to oil, grease, water, mild chemicals, and metal debris
  • Heat-treated king bolt and nut
  • Zinc-plated for corrosion resistance
  • Zerk fittings for easy maintenance
  • Notched yoke base for optional swivel locks
Catalog No. 50-005-27880 Supplier MSC Supplier No. 75948265
This swivel top plate caster features a phenolic wheel with a 6 inch diameter, a 2 inch width, and a 1,200 lb load capacity. Manufactured from cold-formed steel, it includes a heat-treated king bolt and nut for durability. The rig is zinc-plated for corrosion resistance, and zerk fittings in the heat-treated swivel section allow for easy maintenance. It is suitable for material handling and impact-resistant applications.
  • Rolling ease
  • Floor protection
  • Quiet operation
  • Resistance to oil, grease, water, mild chemicals, and metal debris
  • Heat-treated king bolt and nut
  • Zinc-plated for corrosion resistance
  • Zerk fittings for easy maintenance
  • Notched yoke base for optional swivel locks