Learn More
MSC Albion Swivel Top Plate Caster, 6 in Wheel Dia, Polyurethane, 1,000 lb Capacity
Catalog No. 5000527491
Encompass
This swivel top plate caster features a polyurethane wheel with a 6 inch diameter, a 2 inch width, and a load capacity of 1,000 pounds. It has an overall height of 7-1/2 inches and is designed for material handling applications.
  • Swivel section
  • 1/2 in diameter solid axle
  • 1/4 in steel legs
  • Maintenance free
  • Top plate and yoke base
  • Non-marking wheel
  • Cold forged construction
  • Zinc finish/coating
Catalog No. 50-005-27491 Supplier MSC Supplier No. 74877473
This swivel top plate caster features a polyurethane wheel with a 6 inch diameter, a 2 inch width, and a load capacity of 1,000 pounds. It has an overall height of 7-1/2 inches and is designed for material handling applications.
  • Swivel section
  • 1/2 in diameter solid axle
  • 1/4 in steel legs
  • Maintenance free
  • Top plate and yoke base
  • Non-marking wheel
  • Cold forged construction
  • Zinc finish/coating