Learn More
MSC Albion Swivel Top Plate Caster, Phenolic, 8 in Wheel Diameter, 1,250 lb Capacity
Catalog No. 5000512647
Encompass
This swivel top plate caster features a phenolic wheel with an 8-inch diameter and 2-inch width, offering a robust 1,250 lb load capacity and an overall height of 9-1/2 inches. Designed for material handling, it boasts a cold forged swivel construction with a kingpin swivel, zinc finish, and sealed roller bearings for smooth operation.
  • Phenolic wheel, 8 inch diameter, 2 inch width
  • 1,250 lb load capacity
  • Cold forged kingpin swivel construction
  • Zinc finish
  • Sealed roller bearings
  • Top plate and yoke base
  • Nylon seal retaining/thrust washers
  • Zerk fitting
  • Fork legs
Catalog No. 50-005-12647 Supplier MSC Supplier No. 75947507
This swivel top plate caster features a phenolic wheel with an 8-inch diameter and 2-inch width, offering a robust 1,250 lb load capacity and an overall height of 9-1/2 inches. Designed for material handling, it boasts a cold forged swivel construction with a kingpin swivel, zinc finish, and sealed roller bearings for smooth operation.
  • Phenolic wheel, 8 inch diameter, 2 inch width
  • 1,250 lb load capacity
  • Cold forged kingpin swivel construction
  • Zinc finish
  • Sealed roller bearings
  • Top plate and yoke base
  • Nylon seal retaining/thrust washers
  • Zerk fitting
  • Fork legs