Learn More
MSC Albion Swivel Top Plate Caster, Polyurethane, 5 In, 1000 Lb
Catalog No. 5000514202
Encompass
This swivel top plate caster features a polyurethane wheel with a 5 inch diameter and 2 inch width, offering a load capacity of 1000 pounds. It has an overall height of 6-1/2 inches and is designed for robust material handling applications.
  • Roller bearing type.
  • Cold forged construction.
  • Zinc finish.
  • Non-marking.
  • Swivel section.
  • 1/2 in diameter solid axle.
  • 1/4 in steel legs.
  • Maintenance free.
  • Top plate and yoke base.
Catalog No. 50-005-14202 Supplier MSC Supplier No. 74877432
This swivel top plate caster features a polyurethane wheel with a 5 inch diameter and 2 inch width, offering a load capacity of 1000 pounds. It has an overall height of 6-1/2 inches and is designed for robust material handling applications.
  • Roller bearing type.
  • Cold forged construction.
  • Zinc finish.
  • Non-marking.
  • Swivel section.
  • 1/2 in diameter solid axle.
  • 1/4 in steel legs.
  • Maintenance free.
  • Top plate and yoke base.