Learn More
MSC Albion Swivel Top Plate Caster, Polyurethane, 6 inch, 1,230 lb
Catalog No. 5000513086
Encompass
This swivel top plate caster features a durable polyurethane wheel with a 6 inch diameter and a 1,230 lb load capacity, designed for material handling applications. It has a cold forged kingpin swivel construction with a zinc finish, providing reliable performance and maneuverability.
  • Polyurethane wheel with a 6 inch diameter and 2 inch width
  • 1,230 lb load capacity
  • Plate mount for secure attachment
  • Roller bearing for smooth operation
  • Non-marking wheel prevents scuffs on floors
  • Cold forged kingpin swivel construction for durability
  • Features a zerk fitting for easy lubrication
  • Includes nylon seal retaining/thrust washers
  • Zinc finish provides corrosion resistance
  • Designed for material handling applications
  • Overall height of 7-1/4 inch
Catalog No. 50-005-13086 Supplier MSC Supplier No. 75947580
This swivel top plate caster features a durable polyurethane wheel with a 6 inch diameter and a 1,230 lb load capacity, designed for material handling applications. It has a cold forged kingpin swivel construction with a zinc finish, providing reliable performance and maneuverability.
  • Polyurethane wheel with a 6 inch diameter and 2 inch width
  • 1,230 lb load capacity
  • Plate mount for secure attachment
  • Roller bearing for smooth operation
  • Non-marking wheel prevents scuffs on floors
  • Cold forged kingpin swivel construction for durability
  • Features a zerk fitting for easy lubrication
  • Includes nylon seal retaining/thrust washers
  • Zinc finish provides corrosion resistance
  • Designed for material handling applications
  • Overall height of 7-1/4 inch