Learn More
MSC Albion Swivel Top Plate Caster, Rubber, 4 in, 300 lb
Catalog No. 5000530853
Encompass
This swivel top plate caster is designed for institutional and material handling applications. It features a durable rubber wheel, a 300 lb load capacity, and a zinc finish to resist rust and corrosion, ensuring reliable performance.
  • Rubber wheel with 4 in diameter and 1-1/4 in width
  • 300 lb load capacity
  • Overall height of 5-1/8 in
  • Stamp-formed, kingpin swivel design
  • Delrin bearing type for smooth movement
  • Zinc finish/coating to resist rust and corrosion
  • Includes a swivel section and a 3/8 in diameter hollow axle with lock nut
  • Constructed with rubber mold on polypropylene
Catalog No. 50-005-30853 Supplier MSC Supplier No. 75946426
This swivel top plate caster is designed for institutional and material handling applications. It features a durable rubber wheel, a 300 lb load capacity, and a zinc finish to resist rust and corrosion, ensuring reliable performance.
  • Rubber wheel with 4 in diameter and 1-1/4 in width
  • 300 lb load capacity
  • Overall height of 5-1/8 in
  • Stamp-formed, kingpin swivel design
  • Delrin bearing type for smooth movement
  • Zinc finish/coating to resist rust and corrosion
  • Includes a swivel section and a 3/8 in diameter hollow axle with lock nut
  • Constructed with rubber mold on polypropylene