Learn More
MSC Albion Swivel Top Plate Caster, Soft Rubber, 3 in Wheel Diameter, 175 lb Capacity
Catalog No. 5000528943
Encompass
This swivel top plate caster features a soft rubber wheel, a 3 in diameter, and a 175 lb load capacity. It is designed for various material handling applications such as stock trucks.
  • Soft rubber wheel material
  • 3 in wheel diameter
  • 1-1/4 in wheel width
  • 175 lb load capacity
  • Self-lubricating bearing
  • Hardened-heat treated construction
  • Zinc finish
  • 4-3/8 in overall height
  • Ideal for stock trucks and general material handling
Catalog No. 50-005-28943 Supplier MSC Supplier No. 66017229
This swivel top plate caster features a soft rubber wheel, a 3 in diameter, and a 175 lb load capacity. It is designed for various material handling applications such as stock trucks.
  • Soft rubber wheel material
  • 3 in wheel diameter
  • 1-1/4 in wheel width
  • 175 lb load capacity
  • Self-lubricating bearing
  • Hardened-heat treated construction
  • Zinc finish
  • 4-3/8 in overall height
  • Ideal for stock trucks and general material handling