Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hardy Diagnostics™ AlbiQuick™ for Candida Albicans Identification
SDP

Catalog No. 23001761 Shop All Hardy Diagnostics Products
Change view
Click to view available options
Quantity:
25
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-001-761 25
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 23-001-761 Supplier Hardy Diagnostics™ Supplier No. Z121
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

The new AlbiQuick™ utilizes two enzyme color reactions to produce an accurate result that’s easy to read. AlbiQuick is performed on a convenient card with two reaction wells.

  • Only Candida albicans possesses the enzymes that will hydrolyze the PRO and NAG compounds on the card
  • Incubation time is only five minutes at room temperature
  • Requires the use of a handheld 365nm ultra violet lamp
  • The old germ tube method of C. albicans identification requires several hours of incubation and tedious microscopy to produce a result. The AlbiQuick™ eliminates the need for this and other lengthy procedures

Specifications

Description Requires use of a handheld ultraviolet lamp
Quantity 25
Analysis Time 5 min.
Form Solid
Operating Temperature Range 15°C to 30°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.