Learn More
ABclonal Technology Albumin/ALB Rabbit mAb
SDP
Supplier:  ABclonal Technology A24161100UL
Encompass_Preferred
Albumin is a soluble monomeric protein which comprises about one-half of the blood serum protein Albumin functions primarily as a carrier protein for steroids fatty acids and thyroid hormones and plays a role in stabilizing extracellular fluid volume Albumin is a globular unglycosylated serum protein of molecular weight 65 000 Albumin is synthesized in the liver as preproalbumin which has an N-terminal peptide that is removed before the nascent protein is released from the rough endoplasmic reticulum The product proalbumin is in turn cleaved in the Golgi vesicles to produce the secreted albumin
Catalog No. 50-000-06746