Learn More
Active Motif Albumin Blue Fluorescent Assay Kit
SDP
Catalog No. NC9597392
Encompass
Active Motif's Albumin Blue Fluorescent Assay Kit is a quantitative assay designed to measure albumin levels in biological samples including serum and urine. The kit combines unmatched sensitivity, high specificity and a quick, easy protocol to provide an effective assay for studying albumin.
Catalog No. NC9597392 Supplier Active Motif Supplier No. 15002
Active Motif's Albumin Blue Fluorescent Assay Kit is a quantitative assay designed to measure albumin levels in biological samples including serum and urine. The kit combines unmatched sensitivity, high specificity and a quick, easy protocol to provide an effective assay for studying albumin.