Learn More
Crescent Chemical Co Inc ALBUMIN BOVINE 30%
SDP
Supplier:  Crescent Chemical Co Inc SE11937.02
Encompass_Preferred
Albumin bovine, 30 % solution polymer enhanced Higher avidity exclusively controlled through pure albumin polymerisation. Does not contain artificial avidity enhancers or high molecular weight agglutination potentiators (e.g. PVP, gum acacia, or dextran). Does not contain caprylic acid or oth
Catalog No. 50-247-260