Learn More
Crescent Chemical Co Inc ALBUMIN BOVINE CELL CULTURE
SDP
Supplier:  Crescent Chemical Co Inc SE47330.03
Encompass_Preferred
Albumin bovine cell culture grade Cohn Analog. Specially purified albumin with an almost native composition of lipids, fatty acids and other cofactors which are important for cell culture. Suitable as cell growth supplement. Size - 100G Storage Conditio
Catalog No. 50-249-135