Learn More
Crescent Chemical Co Inc ALBUMIN BOVINE CELL CULTURE
SDP
Catalog No. 50249133
Encompass_Preferred
Albumin bovine cell culture grade Cohn Analog. Specially purified albumin with an almost native composition of lipids, fatty acids and other cofactors which are important for cell culture. Suitable as cell growth supplement. Size - 10G Storage Condition
Catalog No. 50-249-133 Supplier Crescent Chemical Co Inc Supplier No. SE47330.01
Albumin bovine cell culture grade Cohn Analog. Specially purified albumin with an almost native composition of lipids, fatty acids and other cofactors which are important for cell culture. Suitable as cell growth supplement. Size - 10G Storage Condition