Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Albumin, Bovine, Cohn Fraction V, 98%, Low Fatty Acid, pH 5.2

Catalog No. p-5386154
Change view
Click to view available options
Quantity:
50 g
100 g
500 g
1 kg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6552518 50 g
AAJ6552522 100 g
AAJ6552536 500 g
AAJ65525A1 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AAJ6552518 Supplier Thermo Scientific Chemicals Supplier No. J6552518
Only null left
Add to Cart

For research use

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Formulation Lyophilized powder
pH Range 5.2
Quantity 50 g
Source Bovine Serum
Storage Requirements 2° to 8°C
Regulatory Status RUO
Product Type Albumin, Bovine, Cohn Fraction V, 98%, Low Fatty Acid, pH 5.2
Recombinant Native
Form Lyophilized
Purity or Quality Grade 98%

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.