Learn More
Crescent Chemical Co Inc ALBUMIN BOVINE CRYST. LYOPHIL.
SDP
Supplier:  Crescent Chemical Co Inc SE11920.06
Encompass_Preferred
Albumin bovine cryst. lyophil. The purest form of our bovine albumins. Manufacturing includes a proprietary heat-shock fractionation process, ion exchange treatment and triple sequential crystallization, resulting in an extremely pure product. Best suited for use
Catalog No. 50-247-238