Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Research Products International Corp Albumin, Bovine Fraction V, 50 G
SDP

Supplier:  Research Products International Corp A3007550.0

Encompass_Preferred

pH (5% water) @ 25°C: 6.5-7.5
Endotoxin: ≤ 3 EU/mg

Molecular Biology Grade.
IgG Free
Low Bioburden
Protease Free
Solvent Free Extraction Process
Manufactured by Heat Shock process

Catalog No. 50-197-8044


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.