Learn More
Crescent Chemical Co Inc ALBUMIN BOVINE FRACTION V
SDP
Supplier:  Crescent Chemical Co Inc SE11946.02
Encompass_Preferred
Albumin bovine Fraction V, pH 7.0 Life Science grade, lyophil. Highly purified albumin ideal for use in biotechnology, cell culture diagnostics, research and other Life Science applications. First isolated by a proprietary heat shock/organic solvent process and further purified by
Catalog No. 50-247-271