Learn More
Crescent Chemical Co Inc ALBUMIN BOVINE FRACTION V
SDP
Catalog No. 50247244
Encompass_Preferred
Albumin bovine Fraction V receptor grade lyophil. Produced by a proprietary heat-shock fractionation with additional solvent treatment. Contains extremely low levels of heavy metals, alkaline earths, fatty acids, and low molecular weight impurities. First choice for diagnos
Catalog No. 50-247-244 Supplier Crescent Chemical Co Inc Supplier No. SE11924.04
Albumin bovine Fraction V receptor grade lyophil. Produced by a proprietary heat-shock fractionation with additional solvent treatment. Contains extremely low levels of heavy metals, alkaline earths, fatty acids, and low molecular weight impurities. First choice for diagnos