Learn More
Crescent Chemical Co Inc ALBUMIN BOVINE FRACTION V
SDP
Supplier:  Crescent Chemical Co Inc SE11945.03
Encompass_Preferred
Albumin bovine Fraction V, Protease and Fatty Acid free diagnostic grade lyophil. Highly purified albumin, especially designed for protease-sensitive immunoassays, such as RIA and EIA. Suitable as protein standard, diluent, enzyme stabilizer. Can be used also in hybridization and nucleic acid based as
Catalog No. 50-247-268