Learn More
Crescent Chemical Co Inc ALBUMIN BOVINE FRACTION V
SDP
Supplier:  Crescent Chemical Co Inc SE11922.03
Encompass_Preferred
Albumin bovine Fraction V, pH 5.2 standard grade, lyophil. Manufactured in a similar way as cat. no. 11930 but with adjustment to pH 5.2 prior to lyophilization. Special quality for serology, antibody enhancement, bacterial and animal culture media. Size - 100G Storage Co
Catalog No. 50-247-240