Learn More
Crescent Chemical Co Inc ALBUMIN BOVINE FRACTION V
SDP
Supplier:  Crescent Chemical Co Inc SE11926.04
Encompass_Preferred
Albumin bovine Fraction V, Protease-free lyophil. Manufactured by a proprietary heat-shock fractionation process; double heated to insure inactivation of proteolytic activity. Excellent for protease sensitive immunoassays, for stabilisation of proteins and enzymes in buffers and before l
Catalog No. 50-247-248