Learn More
Crescent Chemical Co Inc ALBUMIN BOVINE FRACTION V
SDP
Supplier:  Crescent Chemical Co Inc SE11932.02
Encompass_Preferred
Albumin bovine Fraction V, Fatty Acid Free lyophil. Produced by a proprietary heat-shock/solvent fractionation process. Suitable for in vitro binding and transport studies and in diagnostic systems. Size - 25G Storage Conditions - +2 °C TO +8 °C Catalog Number - 11932.02 /
Catalog No. 50-247-254