Learn More
Crescent Chemical Co Inc ALBUMIN BOVINE FRACTION V
SDP
Supplier:  Crescent Chemical Co Inc SE11924.02
Encompass_Preferred
Albumin bovine Fraction V receptor grade lyophil. Produced by a proprietary heat-shock fractionation with additional solvent treatment. Contains extremely low levels of heavy metals, alkaline earths, fatty acids, and low molecular weight impurities. First choice for diagnos
Catalog No. 50-247-242