Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Albumin, Bovine, Fraction V

Catalog No. MP219477205 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP219477205 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP219477205 Supplier MP Biomedicals™ Supplier No. 0219477205
Add to Cart
Add to Cart

Fatty acid free cell culture reagent extracted from bovine.

  • Fraction V, fatty acid free, low endotoxin, cell culture reagent
  • Free Fatty Acids ≤0.05mg/g
  • Endotoxin ≤2 EU/mg
  • Sodium ≤10mg/g
  • Chloride ≤6mg/g
  • CAS: 9048-46-8
TRUSTED_SUSTAINABILITY

Specifications

pH Range 6.8 to 7.2 (7% aq soln)
Quantity 5 g
Storage Requirements +4°C
Product Type Albumin
Species Bovine
Form Flakes

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.