Learn More
Crescent Chemical Co Inc ALBUMIN BOVINE FRACTION V
SDP
Supplier:  Crescent Chemical Co Inc SE11930.04
Encompass_Preferred
Albumin bovine Fraction V, pH 7.0 standard grade, lyophil. Manufactured by a proprietary heat-shock fractionation process, using caprylic acid as an albumin stabilizer. Standard quality for many applications: protein standard, growth promoter in serum-free media for the cultivation
Catalog No. 50-247-252