Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Albumin, Bovine, Fraction V, BAKER™, J.T. Baker™

Catalog No. 02002210 Shop All Avantor Products
Change view
Click to view available options
Quantity:
10 g
Packaging:
Wide Mouth Amber Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
02-002-210 10 g Wide Mouth Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 02-002-210 Supplier Avantor Supplier No. A46402
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

A grade of chemicals for laboratory use

Chemical Identifiers

CAS 9048-46-8
MDL Number MFCD00145743

Specifications

Packaging Wide Mouth Amber Glass Bottle
Quantity 10 g
Chemical Name or Material Albumin, Bovine, Fraction V
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.