Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Albumin, Bovine, Fraction V Powder

Catalog No. MP8810026 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1 kg
5 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP8810026 5 kg
MP08810025 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MP8810026 Supplier MP Biomedicals™ Supplier No. 08810026
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

Albumin is used to solubilize lipids, and is also used as a blocking agent in Western blots or ELISA applications.

Cream, crystalline “flaky” powder. Prior to lyophilization, the product is filtered through a 0.2mm filter. Low ionic strength and bioburden.

  • Solubility: 0.042
  • pH: 7.3 (7% solution in 0.15M NaCl)
  • Purity: 100%
  • Loss on Drying: 3.5%
  • Heavy Metals: <10ppm
  • Additional Tests: Vesicular Stomatitis Virus: Negative; Bluetongue Virus: Negative
  • General: This product was derived from bovine blood collected at a USDA licensed facility. The cattle received ante- and post-mortem health inspection under a veterinarian's supervision and were apparently free from infectious and contagious diseases. All donor animals were sourced from the United States. Veterinary Certificates are maintained on file at the manufacturing site.

TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) Immunoassay, Mammalian Cell Culture
Molecular Weight (g/mol) ∽66
Name Bovine Blood Albumin Fraction V
pH Range 6.8 to 7.2
Preparation Method Prior to lyophilization, the product is filtered through a 0.2μm filter
Quantity 5 kg
Source Bovine Blood
Storage Requirements 2° to 8°C
Form Powder
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.