Learn More
Crescent Chemical Co Inc ALBUMIN BOVINE LOW ENDOTOXIN
SDP
Supplier:  Crescent Chemical Co Inc SE47321.01
Encompass_Preferred
Albumin bovine, Low Endotoxin biotechnology grade Tested negative for mycoplasma and the bovine viruses BVD, IBR, P13, Adeno, Parvo, Rabies, Reo and Bluetongue, in accordance with 9 CFR 113.Suitable as growth promoter in serum-free media for the cultivation of animal cells and
Catalog No. 50-249-127