Learn More
Crescent Chemical Co Inc ALBUMIN BOVINE MOD. COHN
SDP
Supplier:  Crescent Chemical Co Inc SE11943.02
Encompass_Preferred
Albumin bovine, modified Cohn Fraction V, pH 7.0 lyophil. Produced by a modification of the Cohn procedure, including a proprietary heat-shock method and further purification steps by extensive membrane dialysis and filtration. Suitable as protein standard, in ELISA and blotting techniques, as d
Catalog No. 50-247-264